• USA Home
  • Search Results

  • Dpulegone152238982711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(+)-Pulegone

4 Products
Synonym:   (R)-p-Menth-4(8)-en-3-one, (R)-2-Isopropylidene-5-methylcyclohexanone
Image of (+)-Pulegone
CAS Number:
Empirical Formula (Hill Notation):
 C10H16O
Molecular Weight:
 152.23
 Beilstein/REAXYS Number:
 2040703
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Shelf Life
  Special Grade
Show More