• USA Home
  • Search Results

  • Dsodiumlascorbate1981113403211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(+)-Sodium L-ascorbate

4 Products
Synonym:   L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt
Image of (+)-Sodium <SC>L</SC>-ascorbate
CAS Number:
Empirical Formula (Hill Notation):
 C6H7NaO6
Molecular Weight:
 198.11
 Beilstein/REAXYS Number:
 3767246
 EC Number:
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
 eCl@ss:
 34058010
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Loss
  Product Line
  Biological Source
  Special Grade
  Application(s)
  Physical Form
  Color
Show More