• USA Home
  • Search Results

  • Dsparteine2343849208011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(+)-Sparteine

1 Products
Synonym:   Pachycarpine
Image of (+)-Sparteine
CAS Number:
Empirical Formula (Hill Notation):
 C15H26N2
Molecular Weight:
 234.38
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
900263 (+)-Sparteine,