• USA Home
  • Search Results

  • a1antitrypsinfromhumanplasma12345904192311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α1-Antitrypsin from human plasma

2 Products
Synonym:   α1-Proteinase inhibitor
CAS Number:
 EC Number:
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More