• USA Home
  • Search Results

  • aaatrifluorotoluene146119808811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α,α,α-Trifluorotoluene

2 Products
Synonym:   Benzotrifluoride
Image of α,α,α-Trifluorotoluene
CAS Number:
Linear Formula:
 C6H5CF3
Molecular Weight:
 146.11
 Beilstein/REAXYS Number:
 1906908
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More