• USA Home
  • Search Results

  • aadibromomxylene2639662615311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α,α′-Dibromo-m-xylene

1 Products
Synonym:   1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide
Image of α,α′-Dibromo-<I>m</I>-xylene
CAS Number:
Linear Formula:
 C6H4(CH2Br)2
Molecular Weight:
 263.96
 Beilstein/REAXYS Number:
 971085
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More