• USA Home
  • Search Results

  • abamectin123457175141211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Abamectin

1 Products
Synonym:   Avermectin B1
Image of Abamectin
CAS Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More