• USA Home
  • Search Results

  • abirateroneacetate3915515422918211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Abiraterone acetate

3 Products
Synonym:   17-(Pyridin-3-yl)androsta-5,16-dien-3β-yl acetate
Image of Abiraterone acetate
CAS Number:
Empirical Formula (Hill Notation):
 C26H33NO2
Molecular Weight:
 391.55
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More