• USA Home
  • Search Results

  • abscisicacid264321437545211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Abscisic acid

1 Products
Synonym:   (2Z,4E)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-2,4-pentadienoic acid, ABA, Dormin
Image of (±)-Abscisic acid
CAS Number:
Empirical Formula (Hill Notation):
 C15H20O4
Molecular Weight:
 264.32
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More