• USA Home
  • Search Results

  • acacetin2842648044411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acacetin

3 Products
Synonym:   4′-Methylapigenin, 5,7-Dihydroxy-4′-methoxyflavone, Apigenin 4′-methyl ether, Buddleoflavonol, Linarigenin
Image of Acacetin
CAS Number:
Empirical Formula (Hill Notation):
 C16H12O5
Molecular Weight:
 284.26
 Beilstein/REAXYS Number:
 277879
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
 eCl@ss
 32150233
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More