• USA Home
  • Search Results

  • acarotene53687748899511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Carotene

2 Products
Synonym:   (+)-α-Carotene
Image of α-Carotene
CAS Number:
Empirical Formula (Hill Notation):
 C40H56
Molecular Weight:
 536.87
 Beilstein/REAXYS Number:
 3227603
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More