• USA Home
  • Search Results

  • acenaphthened10solution164271506726211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acenaphthene-d10 solution

2 Products
CAS Number:
Empirical Formula (Hill Notation):
 C12D10
Molecular Weight:
 164.27
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Special Grade
  Package quantity
  Concentration
Show More