• USA Home
  • Search Results

  • acenaphthylene1521920896811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acenaphthylene

4 Products
Image of Acenaphthylene
CAS Number:
Empirical Formula (Hill Notation):
 C12H8
Molecular Weight:
 152.19
 Beilstein/REAXYS Number:
 774092
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
  Assay
Show More