• USA Home
  • Search Results

  • acenocoumarol3533315272711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acenocoumarol

1 Products
Synonym:   (±)-Acenocoumarin, 3-(α-Acetonyl-p-nitrobenzyl)-4-hydroxy-coumarin
Image of Acenocoumarol
CAS Number:
Empirical Formula (Hill Notation):
 C19H15NO6
Molecular Weight:
 353.33
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More