• USA Home
  • Search Results

  • acequinocyl384515796019711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acequinocyl

2 Products
Synonym:   2-(Acetyloxy)-3-dodecyl-1,4-naphthalenedione, 3-Acetoxy-2-dodecyl-1,4-naphthoquinone, 3-Dodecyl-1,4-dihydro-1,4-dioxo-2-naphthyl acetate
Image of Acequinocyl
CAS Number:
Empirical Formula (Hill Notation):
 C24H32O4
Molecular Weight:
 384.51
 Beilstein/REAXYS Number:
 2016033
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More