• USA Home
  • Search Results

  • aces18220736582411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

ACES

6 Products
Synonym:   N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
Image of ACES
CAS Number:
Linear Formula:
 H2NCOCH2NHCH2CH2SO3H
Molecular Weight:
 182.20
 Beilstein/REAXYS Number:
 2253770
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  UV-Absorption Values
  Loss
  Solubility
  Product Line
  Special Grade
Show More