• USA Home
  • Search Results

  • acesulfamek201245558962311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acesulfame K

3 Products
Synonym:   6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide potassium salt
Image of Acesulfame K
CAS Number:
Empirical Formula (Hill Notation):
 C4H4KNO4S
Molecular Weight:
 201.24
 Beilstein/REAXYS Number:
 3637857
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Titertest
  Assay
Show More