• USA Home
  • Search Results

  • acetaldehyde44057507011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetaldehyde

11 Products
Synonym:   Ethanal
Image of Acetaldehyde
CAS Number:
Linear Formula:
 CH3CHO
Molecular Weight:
 44.05
 Beilstein/REAXYS Number:
 505984
 Council of Europe no.:
 89
 EC Number:
 FEMA Number:
 2003
 Flavis number:
 5.001
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Suitability
  pH Value
  Vapor pressure
  Transition temperature
  Titertest
  Solubility
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Restriction
  Package quantity
  Document Links
  Allergen
  Organoleptic property
  Biological Source
  Assay
  Special Grade
Show More