• USA Home
  • Search Results

  • acetaminophen1511610390211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetaminophen

5 Products
Synonym:   4′-Hydroxyacetanilide, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, N-Acetyl-4-aminophenol, APAP, Paracetamol
Image of Acetaminophen
CAS Number:
Linear Formula:
 CH3CONHC6H4OH
Molecular Weight:
 151.16
 Beilstein/REAXYS Number:
 2208089
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Assay
  Special Grade
  Application(s)
  Solubility
Show More