• USA Home
  • Search Results

  • acetaminophend4solution155196431536211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetaminophen-D4 solution

2 Products
Image of Acetaminophen-D<SUB>4</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C8D4H5NO2
Molecular Weight:
 155.19
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More