• USA Home
  • Search Results

  • acetanilide1351610384411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetanilide

5 Products
Synonym:   N-Phenylacetamide
Image of Acetanilide
CAS Number:
Linear Formula:
 CH3CONHC6H5
Molecular Weight:
 135.16
 Beilstein/REAXYS Number:
 606468
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39031308
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Purification method
  Melting Point
Show More