• USA Home
  • Search Results

  • acetanisole1501710006111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetanisole

1 Products
Synonym:   4′-Methoxyacetophenone, 4-Acetylanisole, p-Acetanisole
Image of Acetanisole
CAS Number:
Linear Formula:
 CH3OC6H4COCH3
Molecular Weight:
 150.17
 Beilstein/REAXYS Number:
 742313
 Council of Europe no.:
 570
 EC Number:
 FEMA Number:
 2005
 Flavis number:
 7.038
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 39023808
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More