• USA Home
  • Search Results

  • aceticacid10060056419711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetic acid 100%

2 Products
Synonym:   Acetic acid, Ethanoic acid, Glacial acetic acid
Image of Acetic acid 100%
CAS Number:
Linear Formula:
 CH3CO2H
Molecular Weight:
 60.05
 Beilstein/REAXYS Number:
 506007
 EC Index Number:
 200-580-7
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  pH Value
  Viscosity
  Vapor pressure
  Transition temperature
  Autoignition temp
  Assay
Show More