• USA Home
  • Search Results

  • aceticacidd46408118652311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetic acid-d4

3 Products
Synonym:   Acetic-d3 acid-d, Tetradeuteroacetic acid
Image of Acetic acid-d<SUB>4</SUB>
CAS Number:
Linear Formula:
 CD3CO2D
Molecular Weight:
 64.08
 Beilstein/REAXYS Number:
 1748971
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Isotopic Purity
Show More