• USA Home
  • Search Results

  • aceticanhydride1020910824711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetic anhydride

10 Products
Synonym:   Acetyl acetate, Acetyl oxide, Acetyl acetate, Acetyl oxide
Image of Acetic anhydride
CAS Number:
Linear Formula:
 (CH3CO)2O
Molecular Weight:
 102.09
 EC Index Number
 203-564-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  Purity Grade
  Restriction
  Special Grade
  Solubility
Show More