• USA Home
  • Search Results

  • acetoacetamide10110597714011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetoacetamide

2 Products
Synonym:   α-Acetylacetamide, 3-Oxobutanamide
Image of Acetoacetamide
CAS Number:
Linear Formula:
 CH3COCH2CONH2
Molecular Weight:
 101.10
 Beilstein Registry Number:
 1560550
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More