• USA Home
  • Search Results

  • acetochlor269773425682111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetochlor

1 Products
Synonym:   2-Chloro-N-ethoxymethyl-N-(2′-ethyl-6′-methylphenyl) acetamide
Image of Acetochlor
CAS Number:
Empirical Formula (Hill Notation):
 C14H20ClNO2
Molecular Weight:
 269.77
 Beilstein/REAXYS Number:
 2859702
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Shelf Life
Show More