• USA Home
  • Search Results

  • acetone58086764111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetone

32 Products
Synonym:   Dimethyl ketone, Propanone, 2-Propanone
Image of Acetone
CAS Number:
Linear Formula:
 CH3COCH3
Molecular Weight:
 58.08
 Beilstein/REAXYS Number:
 635680
 EC Index Number:
 200-662-2
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Application(s)
  Quality clarifier
  Density
  Anal. Standard Form
  Shelf Life
  Vapor pressure
  Vapor density vs air
  Melting Point
  Explosion limit
  Solubility
  Purity Grade
  Refractive Index
  Ice Shipping
  Agency/Method
  Suitability
  UV-Absorption Condition
  Restriction
  Transmittance
  Capacity
  Autoignition temp
  Physical Form
  Color
  pH Value
  Transition temperature
  Potency
  UV-Absorption Values
  Assay
Show More

Product #
Description
650501 Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%,
270725 Acetone, for HPLC, ≥99.9%,
179124 Acetone, ACS reagent, ≥99.5%,
34850 Acetone, for HPLC, ≥99.8%,
32201 Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC),
320110 Acetone, ACS reagent, ≥99.5%,
439126 Acetone, for HPLC, ≥99.9%,
24201 Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC),
179973 Acetone, Laboratory Reagent, ≥99.5%,
534064 Acetone, histological grade, ≥99.5%,
90872 Acetone, analytical standard,
1.00013 Acetone, EMPROVE® ESSENTIAL Ph Eur,BP,JPE,NF,
W332615 Acetone, natural, ≥97%,
W332607 Acetone, ≥99%, meets FCC analytical specifications,
1.00014 Acetone, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
1.00020 Acetone, for liquid chromatography LiChrosolv®,
1.00012 Acetone, for gas chromatography ECD and FID SupraSolv®,
8.22251 Acetone, EMPLURA®,
1.07021 Acetone, for analysis EMPARTA® ACS,
1.00022 Acetone, for spectroscopy Uvasol®,
1.00658 Acetone, for gas chromatography MS SupraSolv®,
1006801 Acetone, United States Pharmacopeia (USP) Reference Standard,
PHR1219 Acetone, Pharmaceutical Secondary Standard; Certified Reference Material,
AX0120 Acetone, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
809519 Acetone, ACS reagent, ≥99.5%, ≤2 ppm low benzene,
904082 Acetone, 100% Biorenewable, ACS reagent, ≥99.5%, Green Alternative
RTC000078 Acetone, Pharmaceutical Secondary Standard; Certified Reference Material,
AX0115 Acetone, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs,
AX0116 Acetone, OmniSolv®,
AX0110 Acetone, OmniSolv® HR-GC,
AX0125 Acetone, Histology,
AX0114 Acetone, Residue grade,