• USA Home
  • Search Results

  • acetone580898765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetone

32 Products
Image of Acetone
Linear Formula:
 CH3COCH3
Molecular Weight:
 58.08
 Beilstein/REAXYS Number:
 635680
 EC Number:
 MDL number:
 NACRES:
 NA.02
 PubChem Substance ID:
 eCl@ss:
 39021201
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
179124 Acetone, ACS reagent, ≥99.5%,
650501 Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%,
32201 Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC),
270725 Acetone, for HPLC, ≥99.9%,
24201 Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC),
1.00014 Acetone, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
34850 Acetone, for HPLC, ≥99.8%,
179973 Acetone, Laboratory Reagent, ≥99.5%,
90872 Acetone, analytical standard,
1.00012 Acetone, for gas chromatography ECD and FID SupraSolv®,
AX0120 Acetone, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
8.22251 Acetone, EMPLURA®,
PHR1219 Acetone, Pharmaceutical Secondary Standard; Certified Reference Material,
320110 Acetone, ACS reagent, ≥99.5%,
534064 Acetone, histological grade, ≥99.5%,
1.00020 Acetone, for liquid chromatography LiChrosolv®,
439126 Acetone, for HPLC, ≥99.9%,
W332615 Acetone, natural, ≥97%,
1.00013 Acetone, EMPROVE® ESSENTIAL Ph Eur,BP,JPE,NF,
1.00658 Acetone, for gas chromatography MS SupraSolv®,
904082 Acetone, BioRenewable, ACS reagent, ≥99.5%, Green Alternative
AX0115 Acetone, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs,
W332607 Acetone, ≥99%, meets FCC analytical specifications,
1.07021 Acetone, for analysis EMPARTA® ACS,
RTC000078 Acetone, Pharmaceutical Secondary Standard; Certified Reference Material,
1006801 Acetone, United States Pharmacopeia (USP) Reference Standard,
AX0116 Acetone, OmniSolv®,
1.00022 Acetone, for spectroscopy Uvasol®,
AX0125 Acetone, Histology,
AX0114 Acetone, Residue grade,
AX0110 Acetone, OmniSolv® HR-GC,
809519 Acetone, ACS reagent, ≥99.5%, ≤2 ppm low benzene,