• USA Home
  • Search Results

  • acetonecyanohydrin85107586511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetone cyanohydrin

1 Products
Synonym:   α-Hydroxyisobutyronitrile, 2-Hydroxyisobutyronitrile
Image of Acetone cyanohydrin
CAS Number:
Linear Formula:
 (CH3)2C(OH)CN
Molecular Weight:
 85.10
 Beilstein/REAXYS Number:
 605391
 EC Number:
 MDL number:
 NACRES:
 NA.02
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Additive
Show More

Product #
Description
A10000 Acetone cyanohydrin, 99%,