• USA Home
  • Search Results

  • acetoned6641266652411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetone-d6

14 Products
Synonym:   Hexadeuteroacetone
Image of Acetone-d<SUB>6</SUB>
CAS Number:
Linear Formula:
 CD3COCD3
Molecular Weight:
 64.12
 Beilstein/REAXYS Number:
 1702935
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Package quantity
  pH Value
  Transition temperature
  Melting Point
  Explosion limit
  Autoignition temp
  Density
  Vapor pressure
  Potency
  Additive
  Assay
  Isotopic Purity
Show More