• USA Home
  • Search Results

  • acetonitrile41057505811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetonitrile

46 Products
Synonym:   ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide
Image of Acetonitrile
CAS Number:
Linear Formula:
 CH3CN
Molecular Weight:
 41.05
 Beilstein/REAXYS Number:
 741857
 EC Index Number:
 200-835-2
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Shelf Life
  Purification method
  Melting Point
  Compatibility
  Agency/Method
  Suitability
  Purity Grade
  Miscellaneous Specifications
  Measuring range
  Product Line
  Restriction
  UV-Absorption Condition
  Ice Shipping
  Type
  Absorption
  Autoignition temp
  Refractive Index
  Package quantity
  Transmittance
  Physical Form
  Transition temperature
  Vapor pressure
  Solubility
  Color
  Assay
  Special Grade
Show More

Product #
Description
271004 Acetonitrile, anhydrous, 99.8%,
34998 Acetonitrile, HPLC Plus, ≥99.9%,
34851 Acetonitrile, for HPLC, gradient grade, ≥99.9%,
675415 Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles,
360457 Acetonitrile, ACS reagent, ≥99.5%,
270717 Acetonitrile, HPLC grade, ≥99.93%,
439134 Acetonitrile, for HPLC, gradient grade, ≥99.9%,
34881 Acetonitrile, for HPLC-GC, ≥99.8% (GC),
33019 Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC),
110086 Acetonitrile, ReagentPlus®, 99%,
494445 Acetonitrile, biotech. grade, ≥99.93%,
437557 Acetonitrile, ≥99.5%, ACS reagent,
45983 Acetonitrile, analytical standard,
733466 Acetonitrile, electronic grade, 99.999% trace metals basis,
1.13212 Acetonitrile, for DNA synthesis (≤ 30 ppm H₂O),
34442 Acetonitrile, for synthesis of DNA, ≥99.9% (GC),
L011800 Acetonitrile,
1.00030 Acetonitrile, gradient grade for liquid chromatography LiChrosolv® Reag. Ph Eur,
1.14291 Acetonitrile, isocratic grade for liquid chromatography LiChrosolv®,
1.00029 Acetonitrile, hypergrade for LC-MS LiChrosolv®,
AX0142 Acetonitrile, gradient grade OmniSolv®,
AX0145 Acetonitrile, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs,
L010000 Acetonitrile,
1.12636 Acetonitrile, for DNA synthesis (max. 10 ppm H2O),
1.00003 Acetonitrile, for analysis EMSURE® ACS,Reag. Ph Eur,
1.00017 Acetonitrile, for gas chromatography ECD and FID SupraSolv®,
900667 Acetonitrile, for UHPLC, for mass spectrometry,
1.15500 Acetonitrile, EMPLURA®,
L010000 Acetonitrile,
1.00016 Acetonitrile, for spectroscopy Uvasol®,
1.00665 Acetonitrile, for gas chromatography MS SupraSolv®,
L010100 Acetonitrile,
1.13358 Acetonitrile, for preparative chromatography Prepsolv,
L010002 Acetonitrile,
PHR1551 Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material,
1.03725 Acetonitrile, for UHPLC-MS LiChrosolv®,
L010080 Acetonitrile,
L110000 Acetonitrile,
AX0152 Acetonitrile, anhydrous, Biotechnology,
900644 Acetonitrile, anhydrous, ZerO2®, 99.8%,
RTC000080 Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material,
AX0151 Acetonitrile, anhydrous, Biosynthesis OmniSolv®,
AX0143 Acetonitrile, anhydrous, septum-sealed bottle DriSolv,
AX0146 Acetonitrile, Anhydrous,
AX0149 Acetonitrile, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
AX0156 Acetonitrile, OmniSolv® LC-MS,