• USA Home
  • Search Results

  • acetonitrileemproveessential41057505811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetonitrile EMPROVE® ESSENTIAL

1 Products
Synonym:   ACN, Acetonitrile, Cyanomethane, Ethyl nitrile, Methyl cyanide
Image of Acetonitrile EMPROVE<SUP>®</SUP> ESSENTIAL
CAS Number:
Linear Formula:
 CH3CN
Molecular Weight:
 41.05
 Beilstein/REAXYS Number:
 741857
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More