• USA Home
  • Search Results

  • acetonitrilesolution41057505811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetonitrile solution

8 Products
Image of Acetonitrile solution
CAS Number:
Linear Formula:
 CH3CN
Molecular Weight:
 41.05
 Beilstein/REAXYS Number:
 4124158
 EC Number:
 MDL number:
 NACRES:
 NA.03
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Additive
  Vapor pressure
  Vapor density vs air
  Melting Point
  Explosion limit
  Application(s)
Show More

Product #
Description
574732 Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC,
576956 Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC,
900686 Acetonitrile solution, contains 0.1 % (v/v) formic acid, for UHPLC, suitable for mass spectrometry (MS),
574724 Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid,
679186 Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC,
565423 Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC,
703664 Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.,
685461 Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid,