• USA Home
  • Search Results

  • acetonitrilewith005vvtrifluoroaceticacid41057505811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetonitrile with 0.05% (v:v) Trifluoroacetic acid

1 Products
Synonym:   ACN + TFA, Acetonitrile solution
Image of Acetonitrile with 0.05% (v:v) Trifluoroacetic acid
CAS Number:
Linear Formula:
 CH3CN
Molecular Weight:
 41.05
 Beilstein/REAXYS Number:
 4124158
 EC Index Number:
 200-835-2
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More