• USA Home
  • Search Results

  • acetonitrilewith01ammoniumacetate1234514864219711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetonitrile with 0.1% ammonium acetate

1 Products
Image of Acetonitrile with 0.1% ammonium acetate
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.03
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Absorption
Show More