• USA Home
  • Search Results

  • acetonitrilewith01vvtrifluoroaceticacid41057505811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetonitrile with 0.1% (v:v) trifluoroacetic acid

2 Products
Synonym:   ACN + TFA, Acetonitrile solution
Image of Acetonitrile with 0.1% (v:v) trifluoroacetic acid
CAS Number:
Linear Formula:
 CH3CN
Molecular Weight:
 41.05
 Beilstein/REAXYS Number:
 4124158
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Vapor pressure
  Transition temperature
Show More