• USA Home
  • Search Results

  • acetophenone120159886211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetophenone

7 Products
Synonym:   Methyl phenyl ketone
Image of Acetophenone
CAS Number:
Linear Formula:
 CH3COC6H5
Molecular Weight:
 120.15
 Beilstein/REAXYS Number:
 605842
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Document Links
  Anal. Standard Form
  Shelf Life
  Allergen
  Organoleptic property
  Transition temperature
  Purity Grade
  Potency
  Biological Source
  Special Grade
  Refractive Index
  Application(s)
  Assay
  Physical Form
Show More