• USA Home
  • Search Results

  • acetylacetone1001212354611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetylacetone

5 Products
Synonym:   2,4-Pentanedione
Image of Acetylacetone
CAS Number:
Linear Formula:
 CH3COCH2COCH3
Molecular Weight:
 100.12
 Beilstein/REAXYS Number:
 741937
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39021208
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Transition temperature
  Application(s)
  Potency
  Special Grade
  Physical Form
  pH Value
  Assay
Show More