• USA Home
  • Search Results

  • acetylbcyclodextrin1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetyl-β-cyclodextrin

1 Products
Image of Acetyl-β-cyclodextrin
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More