• USA Home
  • Search Results

  • acetylcholinechloride181666031111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetylcholine chloride

6 Products
Synonym:   ACh
Image of Acetylcholine chloride
CAS Number:
Linear Formula:
 (CH3)3N+CH2CH2OCOCH3Cl-
Molecular Weight:
 181.66
 Beilstein/REAXYS Number:
 3571875
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Special Grade
  Solubility
  Package quantity
  Anal. Standard Form
Show More