• USA Home
  • Search Results

  • acetylcholineiodide27311226050611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetylcholine iodide

2 Products
Synonym:   ACh
Image of Acetylcholine iodide
CAS Number:
Linear Formula:
 (CH3)3N(I)CH2CH2OCOCH3
Molecular Weight:
 273.11
 Beilstein/REAXYS Number:
 3571339
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More