• USA Home
  • Search Results

  • acetylcoenzymeasodiumsalt8095710202973211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetyl coenzyme A sodium salt

1 Products
Synonym:   Acetyl CoA, Acetyl-S- CoA
Image of Acetyl coenzyme A sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C23H38N7O17P3S
Molecular Weight:
 809.57
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More