• USA Home
  • Search Results

  • acetylpyrazine122122204725211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acetylpyrazine

1 Products
Image of Acetylpyrazine
CAS Number:
Empirical Formula (Hill Notation):
 C6H6N2O
Molecular Weight:
 122.12
 Beilstein/REAXYS Number:
 109630
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
251801 Acetylpyrazine, 97%,