• USA Home
  • Search Results

  • achloralose309531587993311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Chloralose

2 Products
Synonym:   1,2-O-(2,2,2-Trichloroethylidene)-α-D-glucofuranose, Anhydro-D-glucochloral, Chloralose
Image of α-Chloralose
CAS Number:
Empirical Formula (Hill Notation):
 C8H11Cl3O6
Molecular Weight:
 309.53
 Beilstein/REAXYS Number:
 85418
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More