• USA Home
  • Search Results

  • acibenzolaracid180183527227611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acibenzolar acid

1 Products
Synonym:   1,2,3-Benzothiadiazole-7-carboxylic acid
Image of Acibenzolar acid
CAS Number:
Empirical Formula (Hill Notation):
 C7H4N2O2S
Molecular Weight:
 180.18
 Beilstein/REAXYS Number:
 975928
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More