• USA Home
  • Search Results

  • acibenzolarsmethyl2102813515854211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acibenzolar-S-methyl

2 Products
Synonym:   S-Methyl-1,2,3-benzothiadiazole-7-carbothioate
Image of Acibenzolar-<I>S</I>-methyl
CAS Number:
Empirical Formula (Hill Notation):
 C8H6N2OS2
Molecular Weight:
 210.28
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More