• USA Home
  • Search Results

  • aciclovirforsystemsuitability225205927789311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aciclovir for system suitability

1 Products
Synonym:   9-[(2-Hydroxyethoxy)methyl]guanine, Acycloguanosine, Acyclovir
Image of Aciclovir for system suitability
CAS Number:
Empirical Formula (Hill Notation):
 C8H11N5O3
Molecular Weight:
 225.20
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More