• USA Home
  • Search Results

  • acidred7355648541375211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acid Red 73

1 Products
Synonym:   7-Hydroxy-8-(4-phenylazophenylazo)-1,3-naphthalenedisulfonic acid disodium salt, Acid Red 73, Brilliant Crocein MOO, Brilliant crocein scarlet MOO, Crocein Scarlet MOO
Image of Acid Red 73
CAS Number:
Empirical Formula (Hill Notation):
 C22H14N4Na2O7S2
Molecular Weight:
 556.48
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More