• USA Home
  • Search Results

  • acifluorfen361665059466611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Acifluorfen

2 Products
Synonym:   5-(2-Chloro-α,α,α-trifluoro-p-tolyloxy)-2-nitrobenzoic acid, 5-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoic acid
Image of Acifluorfen
CAS Number:
Empirical Formula (Hill Notation):
 C14H7ClF3NO5
Molecular Weight:
 361.66
 Beilstein/REAXYS Number:
 2953865
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
Show More